Gadolinium, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

693723-1EA
SIAL693723-1EAEA 367 EUR
SIAL693723-1EA
Gadolinium, Sigma-Aldrich®
Gadolinium
Formel: Gd
Molecular Weight: 157,25 g/mol
MDL: MFCD00011022
CAS-Nummer: 7440-54-2
EINECS: 231-162-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR