2-(Benzyloxy)benzoesäure, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2-Benzyloxybenzoic acid

M-Clarity™ Qualitätsstufe = MQ100

693995-1G
SIAL693995-1GEA 70.4 EUR
SIAL693995-1G
2-(Benzyloxy)benzoesäure, Sigma-Aldrich®
2-(Benzyloxy)benzoesäure
Formel: C₁₄H₁₂O₃
Molecular Weight: 228,25 g/mol
MDL: MFCD00051940
CAS-Nummer: 14389-86-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR