3-Methylbenzo[b]thiophen-2-carbaldehyd, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

694320-1G
SIAL694320-1GEA 47.5 EUR
SIAL694320-1G
3-Methylbenzo[b]thiophen-2-carbaldehyd, Sigma-Aldrich®
3-Methylbenzo[b]thiophen-2-carbaldehyd
Formel: C₁₀H₈OS
Molecular Weight: 176,24 g/mol
MDL: MFCD00052307
CAS-Nummer: 22053-74-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR