Methyl-1-trifluormethyl-1-cyclopropancarboxylat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Methyl 1-(trifluoromethyl)cyclopropanecarboxylate

M-Clarity™ Qualitätsstufe = MQ100

694363-1G
SIAL694363-1GEA 236 EUR
SIAL694363-1G
Methyl-1-trifluormethyl-1-cyclopropancarboxylat, Sigma-Aldrich®
Methyl-1-trifluormethyl-1-cyclopropancarboxylat
Formel: C₆H₇F₃O₂
Molecular Weight: 168,12 g/mol
MDL: MFCD06801321
CAS-Nummer: 208242-25-5
UN: 3272
ADR: 3,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR