2-Fluorcyclohexanon, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

694533-1G
SIAL694533-1GEA 182 EUR
SIAL694533-1G
2-Fluorcyclohexanon, Sigma-Aldrich®
2-Fluorocyclohexanone
Formel: C₆H₉FO
MDL: MFCD01076476
CAS-Nummer: 694-82-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR