(3,3,3-Trifluorpropyl)dimethoxymethylsilan, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Dimethoxy(methyl)(3,3,3-trifluoropropyl)silane, 1,1,1-Trifluoro-3-[dimethoxy(methyl)silyl]propane

69465-2.5ML
SIAL69465-2.5MLEA 239 EUR
SIAL69465-2.5ML
(3,3,3-Trifluorpropyl)dimethoxymethylsilan, Sigma-Aldrich®
(3,3,3-Trifluorpropyl)dimethoxymethylsilan
Formel: C₆H₁₃F₃O₂Si
Molecular Weight: 202,25 g/mol
MDL: MFCD00053175
CAS-Nummer: 358-67-8
EINECS: 206-619-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR