4-Fluor-1-naphthaldehyd, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

694886-1G
SIAL694886-1GEA 113 EUR
SIAL694886-1G
4-Fluor-1-naphthaldehyd, Sigma-Aldrich®
4-Fluor-1-naphthaldehyd
Formel: C₁₁H₇FO
Molecular Weight: 174,1700 g/mol
MDL: MFCD02261765
CAS-Nummer: 172033-73-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR