1,1'-Bis(di-tert-butylphosphino)ferrocen, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

695149-1G
SIAL695149-1GEA 149 EUR
SIAL695149-1G
1,1'-Bis(di-tert-butylphosphino)ferrocen, Sigma-Aldrich®
1,1'-Bis(di-tert-butylphosphino)ferrocen
Formel: C₂₆H₄₄FeP₂
MDL: MFCD01630818
CAS-Nummer: 84680-95-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR