Chlorbis(4-methylphenyl)phosphin, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Chlorodi(p-tolyl)phosphine

M-Clarity™ Qualitätsstufe = MQ100

695815-1G
SIAL695815-1GEA 94 EUR
SIAL695815-1G
Chlorbis(4-methylphenyl)phosphin, Sigma-Aldrich®
Chlorbis(4-methylphenyl)phosphin
Formel: C₁₄H₁₄ClP
Molecular Weight: 248,69 g/mol
MDL: MFCD01630844
CAS-Nummer: 1019-71-2
UN: 1760
ADR: 8,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR