2-Cyan-4-methylanilin, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2-Amino-5-methylbenzonitril, 2-Amino-5-methylbenzoesäurenitril

M-Clarity™ Qualitätsstufe = MQ100

696331-1G
SIAL696331-1GEA 37.6 EUR
SIAL696331-1G
2-Cyan-4-methylanilin, Sigma-Aldrich®
2-Cyan-4-methylanilin
Formel: C₈H₈N₂
Molecular Weight: 132,16 g/mol
MDL: MFCD00267506
CAS-Nummer: 5925-93-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR