5-Hydroxy-o-toluylsäure, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 5-Hydroxy-2-methylbenzoic acid

M-Clarity™ Qualitätsstufe = MQ100

696366-1G
SIAL696366-1GEA 34.8 EUR
SIAL696366-1G
5-Hydroxy-o-toluylsäure, Sigma-Aldrich®
5-Hydroxy-o-toluylsäure
Formel: C₈H₈O₃
Molecular Weight: 152,15 g/mol
MDL: MFCD09833184
CAS-Nummer: 578-22-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR