(S)-(+)-Ethyl-3-piperidincarboxylat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Ethyl L-nipecotate, Ethyl (S)-(+)-3-Piperidinecarboxylate, (S)-Ethyl piperidine-3-carboxylate

M-Clarity™ Qualitätsstufe = MQ100

696439-1G
SIAL696439-1GEA 40.6 EUR
SIAL696439-1G
(S)-(+)-Ethyl-3-piperidincarboxylat, Sigma-Aldrich®
(S)-(+)-Ethyl-3-piperidincarboxylat
Formel: C₈H₁₅NO₂
Molecular Weight: 157,21 g/mol
MDL: MFCD00792499
CAS-Nummer: 37675-18-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR