1-Boc-4-cyanpiperidin, Sigma-Aldrich®

Lieferant: Merck

Synonyme: tert-Butyl 4-cyanopiperidine-1-carboxylate, N-BOC-Piperidine-4-carbonitrile

M-Clarity™ Qualitätsstufe = MQ100

696447-1G
SIAL696447-1GEA 46.2 EUR
SIAL696447-1G
1-Boc-4-cyanpiperidin, Sigma-Aldrich®
1-Boc-4-cyanpiperidin
Formel: C₁₁H₁₈N₂O₂
Molecular Weight: 210,27 g/mol
MDL: MFCD01861223
CAS-Nummer: 91419-52-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR