Ethyl-4-oxazolcarboxylat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Ethyl oxazole-4-carboxylate

696625-1G
SIAL696625-1GEA 70.6 EUR
SIAL696625-1G
Ethyl-4-oxazolcarboxylat, Sigma-Aldrich®
Ethyl-4-oxazolcarboxylat
Formel: C₆H₇NO₃
Molecular Weight: 141,1300 g/mol
MDL: MFCD04114940
CAS-Nummer: 23012-14-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR