8-Fluor-4-chinolinol, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 8-Fluoroquinolin-4-ol, 8-Fluoro-4-hydroxyquinoline

M-Clarity™ Qualitätsstufe = MQ100

697052-1G
SIAL697052-1GEA 69.6 EUR
SIAL697052-1G
8-Fluor-4-chinolinol, Sigma-Aldrich®
8-Fluor-4-chinolinol
Formel: C₉H₆FNO
Molecular Weight: 163,15 g/mol
MDL: MFCD00272395
CAS-Nummer: 63010-71-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR