2,6-Diamino-3,5-difluorpyridin, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

697087-1G
SIAL697087-1GEA 34.5 EUR
SIAL697087-1G
2,6-Diamino-3,5-difluorpyridin, Sigma-Aldrich®
2,6-Diamino-3,5-difluoropyridine
Formel: C₅H₅F₂N₃
Molecular Weight: 145,11 g/mol
MDL: MFCD03412208
CAS-Nummer: 247069-27-8
UN: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR