2-Chlor-3-pyridinboronsäure, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2-Chlor-3-pyridinylboronsäure, 2-Chlorpyridin-3-boronsäure

M-Clarity™ Qualitätsstufe = MQ100

697095-1G
SIAL697095-1GEA 48.2 EUR
SIAL697095-1G
2-Chlor-3-pyridinboronsäure, Sigma-Aldrich®
2-Chloro-3-pyridineboronic acid
Formel: C₅H₅BClNO₂
Molecular Weight: 157,36 g/mol
MDL: MFCD03094997
CAS-Nummer: 381248-04-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR