6-Brom-m-toluonitril, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

697168-1G
SIAL697168-1GEA 29.7 EUR
SIAL697168-1G
6-Brom-m-toluonitril, Sigma-Aldrich®
6-Brom-m-toluonitril
Formel: C₈H₆BrN
Molecular Weight: 196,05 g/mol
MDL: MFCD11040281
CAS-Nummer: 42872-83-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR