3-Brom-2-hydroxybenzonitril, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

697192-1G
SIAL697192-1GEA 86.3 EUR
SIAL697192-1G
3-Brom-2-hydroxybenzonitril, Sigma-Aldrich®
3-Brom-2-hydroxybenzonitril
Formel: C₇H₄BrNO
Molecular Weight: 198,02 g/mol
MDL: MFCD11042854
CAS-Nummer: 13073-28-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR