Bis(2,2,6,6-tetramethyl-3,5-heptandionato-O,O')strontium, Sigma-Aldrich®

Synonyme: Bis(2,2,6,6-tetramethyl-3,5-heptanedionato)strontium(II)

697524-1G
SIAL697524-1GEA 68.2 EUR
SIAL697524-1G
Bis(2,2,6,6-tetramethyl-3,5-heptandionato-O,O')strontium, Sigma-Aldrich®
Bis(2,2,6,6-tetramethyl-3,5-heptandionato-O,O')strontium
Formel: C₂₂H₃₈O₄Sr
MDL: MFCD00064758
CAS-Nummer: 36830-74-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR