Methyl-2-fluorisonicotinat, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
Methyl 2-fluoroisonicotinate
M-Clarity™ Qualitätsstufe = MQ100
697869-250MG
SIAL697869-250MGEA
29.4
EUR
SIAL697869-250MG
Methyl-2-fluorisonicotinat, Sigma-Aldrich®
Methyl-2-fluorisonicotinat
Formel:
C₇H₆FNO₂ Molecular Weight: 155,1300 g/mol |
MDL:
MFCD11656063 CAS-Nummer: 455-69-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...