6-Bromchinolin, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

697893-5G
SIAL697893-5GEA 108 EUR
SIAL697893-5G
6-Bromchinolin, Sigma-Aldrich®
6-Bromoquinoline
Formel: C₉H₆BrN
Molecular Weight: 208,06 g/mol
MDL: MFCD00024023
CAS-Nummer: 5332-25-2
EINECS: 226-238-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR