5,5-Dimethylacetylaceton, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 5,5-Dimethylhexane-2,4-dione, 2,2-Dimethyl-3,5-hexanedione, 5,5-Diméthylhexanedione-2,4

M-Clarity™ Qualitätsstufe = MQ100

697958-1G
SIAL697958-1GEA 43.1 EUR
SIAL697958-1G
5,5-Dimethylacetylaceton, Sigma-Aldrich®
5,5-Dimethylacetylaceton
Formel: C₈H₁₄O₂
Molecular Weight: 142,2 g/mol
MDL: MFCD00142767
CAS-Nummer: 7307-04-2
EINECS: 230-760-0
UN: 1224
ADR: 3,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR