1,2-Bis(diethylphosphino)ethan, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

698296-500MG
SIAL698296-500MGEA 77.8 EUR
SIAL698296-500MG
1,2-Bis(diethylphosphino)ethan, Sigma-Aldrich®
1,2-Bis(diethylphosphino)ethan
Formel: C₁₀H₂₄P₂
MDL: MFCD00015175
CAS-Nummer: 6411-21-8
UN: 2845
ADR: 4.2,I

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR