Chlorbis(4-methoxyphenyl)phosphin, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

698474-250MG
SIAL698474-250MGEA 80.7 EUR
SIAL698474-250MG
Chlorbis(4-methoxyphenyl)phosphin, Sigma-Aldrich®
Chlorbis(4-methoxyphenyl)phosphin
Formel: C₁₄H₁₄ClO₂P
Molecular Weight: 280,69 g/mol
MDL: MFCD04972301
CAS-Nummer: 13685-30-8
UN: 3261
ADR: 8,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR