1,2-Bis(dichlorphosphino)benzol, SAFC®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

698490-250MG
SIAL698490-250MGEA 0 EUR
SIAL698490-250MG
1,2-Bis(dichlorphosphino)benzol, SAFC®
1,2-Bis(dichlorphosphino)benzol
Formel: C₆H₄Cl₄P₂
CAS-Nummer: 82495-67-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR