1-Methyl-1H-pyrazol-5-boronsäurepinakolester, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 1-Methyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrazole, 4,4,5,5-Tetramethyl-2-(1-methylpyrazol-5-yl)-1,3,2-dioxaborolane, 1-Methyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole, 1-Methylpyrazole-5-boronic acid Pinacol ester

M-Clarity™ Qualitätsstufe = MQ100

698628-5G
SIAL698628-5GEA 52.9 EUR
SIAL698628-5G
1-Methyl-1H-pyrazol-5-boronsäurepinakolester, Sigma-Aldrich®
1-Methyl-1H-pyrazol-5-boronsäurepinakolester
Formel: C₁₀H₁₇BN₂O₂
Molecular Weight: 208.07 g/mol
MDL: MFCD05861380
CAS-Nummer: 847818-74-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR