2-Formyl-3-picolin, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 3-Methylpyridine-2-carboxaldehyde, 3-Methylpicolinaldehyde

699071-1G
SIAL699071-1GEA 138 EUR
SIAL699071-1G
2-Formyl-3-picolin, Sigma-Aldrich®
2-Formyl-3-picolin
Formel: C₇H₇NO
Molecular Weight: 121,14 g/mol
MDL: MFCD02181144
CAS-Nummer: 55589-47-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR