3-(4-Morpholinylcarbonyl)phenylboronsäurepinakolester, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Morpholino(3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)methanone, 3-(4-Morpholinylcarbonyl)benzeneboronic acid pinacol ester

M-Clarity™ Qualitätsstufe = MQ100

699438-5G
SIAL699438-5GEA 169 EUR
SIAL699438-5G
3-(4-Morpholinylcarbonyl)phenylboronsäurepinakolester, Sigma-Aldrich®
3-(4-Morpholinylcarbonyl)phenylboronsäurepinakolester
Formel: C₁₇H₂₄BNO₄
Molecular Weight: 317.19 g/mol
MDL: MFCD09266172
CAS-Nummer: 1036991-25-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR