3-(Morpholinomethyl)phenylboronsäurepinakolester, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 4-(3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzyl)morpholine, 3-(4-Morpholinylmethyl)benzeneboronic acid pinacol ester

M-Clarity™ Qualitätsstufe = MQ100

699446-1G
SIAL699446-1GEA 22.1 EUR
SIAL699446-1G
3-(Morpholinomethyl)phenylboronsäurepinakolester, Sigma-Aldrich®
3-(Morpholinomethyl)phenylboronsäurepinakolester
Formel: C₁₇H₂₆BNO₃
Molecular Weight: 303,20 g/mol
MDL: MFCD06797489
CAS-Nummer: 364794-80-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR