3-(4-Fluorphenyl)-5-methyl-4-isoxazolcarbonsäure, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 3-(4-Fluorophenyl)-5-methylisoxazol-4-carboxylic acid

M-Clarity™ Qualitätsstufe = MQ100

699942-1G
SIAL699942-1GEA 77 EUR
SIAL699942-1G
3-(4-Fluorphenyl)-5-methyl-4-isoxazolcarbonsäure, Sigma-Aldrich®
3-(4-Fluorophenyl)-5-methylisoxazole-4-carboxylic acid
Formel: C₁₁H₈FNO₃
Molecular Weight: 221,19 g/mol
MDL: MFCD03407356
CAS-Nummer: 1736-21-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR