2-4-Bromphenyl)chinoxalin, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

700061-5G
SIAL700061-5GEA 80.9 EUR
SIAL700061-5G
2-4-Bromphenyl)chinoxalin, Sigma-Aldrich®
2-4-Bromphenyl)chinoxalin
Formel: C₁₄H₉BrN₂
MDL: MFCD00222258
CAS-Nummer: 5021-45-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR