2,3-Dibrom-4-methylthiophen, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

700282-1G
SIAL700282-1GEA 211 EUR
SIAL700282-1G
2,3-Dibrom-4-methylthiophen, Sigma-Aldrich®
2,3-Dibromo-4-methylthiophene
Formel: C₅H₄Br₂S
CAS-Nummer: 125257-38-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR