2-Chlor-6-methoxyisonicotinsäure, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2-Chloro-6-methoxypyridine-4-carboxylic acid

M-Clarity™ Qualitätsstufe = MQ100

700614-1G
SIAL700614-1GEA 89.3 EUR
SIAL700614-1G
2-Chlor-6-methoxyisonicotinsäure, Sigma-Aldrich®
2-Chlor-6-methoxyisonicotinsäure
Formel: C₇H₆ClNO₃
Molecular Weight: 187,58 g/mol
MDL: MFCD00173928
CAS-Nummer: 15855-06-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR