5-(Trifluormethyl)picolinsäure, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 5-(Trifluoromethyl)pyridine-2-carboxylic acid

M-Clarity™ Qualitätsstufe = MQ100

700630-500MG
SIAL700630-500MGEA 136 EUR
SIAL700630-500MG
5-(Trifluormethyl)picolinsäure, Sigma-Aldrich®
5-(Trifluormethyl)picolinsäure
Formel: C₇H₄F₃NO₂
Molecular Weight: 191,11 g/mol
MDL: MFCD04113632
CAS-Nummer: 80194-69-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR