5-Brom-4,6-difluorpyrimidin, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

700975-250MG
SIAL700975-250MGEA 140 EUR
SIAL700975-250MG
5-Brom-4,6-difluorpyrimidin, Sigma-Aldrich®
5-Bromo-4,6-difluoropyrimidine
Formel: C₄HBrF₂N₂
Molecular Weight: 194,9650 g/mol
MDL: MFCD08686978
CAS-Nummer: 946681-88-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR