5-Fluor-1H-indazol, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

701025-500MG
SIAL701025-500MGEA 134 EUR
SIAL701025-500MG
5-Fluor-1H-indazol, Sigma-Aldrich®
5-Fluoro-1H-indazole
Formel: C₇H₅FN₂
Molecular Weight: 136,13 g/mol
MDL: MFCD04972877
CAS-Nummer: 348-26-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR