2-Amino-3,5-difluor-4-(trifluormethyl)pyridin, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

701033-250MG
SIAL701033-250MGEA 142 EUR
SIAL701033-250MG
2-Amino-3,5-difluor-4-(trifluormethyl)pyridin, Sigma-Aldrich®
2-Amino-3,5-difluoro-4-(trifluoromethyl)pyridine
Formel: C₆H₃F₅N₂
MDL: MFCD03840199
CAS-Nummer: 883498-68-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR