3-Brom-4-hydroxy-5-methoxybenzonitril, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

701165-5G
SIAL701165-5GEA 135 EUR
SIAL701165-5G
3-Brom-4-hydroxy-5-methoxybenzonitril, Sigma-Aldrich®
3-Brom-4-hydroxy-5-methoxybenzonitril
Formel: C₈H₆BrNO₂
Molecular Weight: 228,0400 g/mol
MDL: MFCD01123027
CAS-Nummer: 52805-45-5
UN: 3439
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR