3-Brom-2-methoxybenzonitril, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 3-Brom-2-methoxybenzoesäurenitril

M-Clarity™ Qualitätsstufe = MQ100

701181-1G
SIAL701181-1GEA 49.2 EUR
SIAL701181-1G
3-Brom-2-methoxybenzonitril, Sigma-Aldrich®
3-Brom-2-methoxybenzonitril
Formel: C₈H₆BrNO
Molecular Weight: 212,05 g/mol
MDL: MFCD09834773
CAS-Nummer: 874472-98-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR