2-Brom-5-nitrobenzonitril, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 1-Cyano-2-bromo-5-nitrobenzene

M-Clarity™ Qualitätsstufe = MQ100

701238-5G
SIAL701238-5GEA 66.8 EUR
SIAL701238-5G
2-Brom-5-nitrobenzonitril, Sigma-Aldrich®
2-Brom-5-nitrobenzonitril
Formel: C₇H₃BrN₂O₂
Molecular Weight: 227,02 g/mol
MDL: MFCD00234249
CAS-Nummer: 134604-07-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR