Methyl-2-fluornicotinat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Methyl 2-fluoronicotinate, 2-Fluoronicotinic acid methyl ester

M-Clarity™ Qualitätsstufe = MQ100

701904-1G
SIAL701904-1GEA 96.8 EUR
SIAL701904-1G
Methyl-2-fluornicotinat, Sigma-Aldrich®
Methyl-2-fluornicotinat
Formel: C₇H₆FNO₂
Molecular Weight: 155,13 g/mol
MDL: MFCD03095072
CAS-Nummer: 446-26-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR