Carbon black, mesoporös, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Furnace black, Lamp Black

M-Clarity™ Qualitätsstufe = MQ100

702110-5G
SIAL702110-5GEA 286 EUR
SIAL702110-5G
Carbon black, mesoporös, Sigma-Aldrich®
Carbon black

Carbon with hydrophilic pore surface.


  • Particle size: 5 to 50 μm (SEM)
  • Pore size: >0.2 cm³/g mesoporosity
  • Specific surface area: >50 m²/g (BET)


Starbons: Cooking up nanostructured mesoporous materials.

Formel: C
Molecular Weight: 12,01 g/mol
Siedepunkt: 4827 °C
Schmelzpunkt: 3654… 3697 °C
MDL: MFCD00133992
CAS-Nummer: 1333-86-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR