5-Brom-1H-indazol, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

702293-1G
SIAL702293-1GEA 41.8 EUR
SIAL702293-1G
5-Brom-1H-indazol, Sigma-Aldrich®
5-Bromo-1H-indazole
Formel: C₇H₅BrN₂
Molecular Weight: 197,03 g/mol
MDL: MFCD00839493
CAS-Nummer: 53857-57-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR