Dinatriumnaphthalin-1,5-disulfonat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 1,5-Naphthalindisulfonsäure Dinatriumsalz, Naphthalin-1,5-disulfonsäure Dinatriumsalz

70240-1KG
SIAL70240-1KGEA 123 EUR
SIAL70240-1KG
Dinatriumnaphthalin-1,5-disulfonat, Sigma-Aldrich®
Dinatriumnaphthalin-1,5-disulfonat
Formel: C₁₀H₆Na₂O₆S₂
Molecular Weight: 332.27 g/mol
MDL: MFCD00064178
CAS-Nummer: 1655-29-4
EINECS: 216-732-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR