1H-Indazol-6-carbonsäure, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

702420-1G
SIAL702420-1GEA 94.9 EUR
SIAL702420-1G
1H-Indazol-6-carbonsäure, Sigma-Aldrich®
1H-Indazole-6-carboxylic acid
Formel: C₈H₆N₂O₂
Molecular Weight: 162,1454 g/mol
MDL: MFCD06804571
CAS-Nummer: 704-91-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR