4-Methyl-2-(2-pyridinyl)-5-thiazolcarbonsäure, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

702528-1G
SIAL702528-1GEA 48.1 EUR
SIAL702528-1G
4-Methyl-2-(2-pyridinyl)-5-thiazolcarbonsäure, Sigma-Aldrich®
4-Methyl-2-(2-pyridinyl)-5-thiazolcarbonsäure
Formel: C₁₀H₈N₂O₂S
Molecular Weight: 220,25 g/mol
MDL: MFCD01316364
CAS-Nummer: 34418-48-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR