4-Methyl-2-(4-pyridinyl)-5-thiazolcarbonsäure, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 4-Methyl-2-(pyridin-4-yl)thiazole-5-carboxylic acid

M-Clarity™ Qualitätsstufe = MQ100

702536-1G
SIAL702536-1GEA 97 EUR
SIAL702536-1G
4-Methyl-2-(4-pyridinyl)-5-thiazolcarbonsäure, Sigma-Aldrich®
4-Methyl-2-(4-pyridinyl)-5-thiazolcarbonsäure
Formel: C₁₀H₈N₂O₂S
Molecular Weight: 220,2477 g/mol
MDL: MFCD00171757
CAS-Nummer: 144060-98-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR