Kaliumbis(trimethylsilyl)amid 1 mol/l in THF, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ200

702722-100ML
SIAL702722-100MLEA 194 EUR
SIAL702722-100ML
Kaliumbis(trimethylsilyl)amid 1 mol/l in THF, Sigma-Aldrich®
Kaliumbis(trimethylsilyl)amid
Formel: C₆H₁₈KNSi₂
MDL: MFCD00010330
CAS-Nummer: 40949-94-8
EINECS: 424-100-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR