Kupfer(II)phthalocyanin, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
[29H,31H-Phthalocyaninato-N29,N30,N31,N32]Kupfer, CuPc, Phthalocyanin-Kupfer(II)-Komplex, Phthalocyaninblau, Tetrabenzo-5,10,15,20-diazaporphyrinphthalocyanin, Pigment Blue 15, Kupfer-Phthalocyanin, Heliogenblau B
M-Clarity™ Qualitätsstufe = MQ100
702854-500MG
SIAL702854-500MGEA
308
EUR
SIAL702854-500MG
Kupfer(II)phthalocyanin, Sigma-Aldrich®
Copper(II) phthalocyanine
Formel:
C₃₂H₁₆CuN₈ Molecular Weight: 576,08 g/mol Schmelzpunkt: 480 °C |
MDL:
MFCD00010719 CAS-Nummer: 147-14-8 EINECS: 205-685-1 Merck Index: 12,02584 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...